missing translation for 'onlineSavingsMsg'
Learn More

4-Methyloxazole-5-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7133358
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
200 mg
Catalog No. Quantity
M2663200MG 200 mg
M26631G 1 g
2 options

Catalog No. M2663200MG

Supplier: TCI America M2663200MG

Only null left

Chemical Identifiers

CAS 2510-32-9
Molecular Formula C5H5NO3
Molecular Weight (g/mol) 127.099
MDL Number MFCD01571070
InChI Key ZIXUNDOOBLSXPE-UHFFFAOYSA-N
Synonym 4-methyloxazole-5-carboxylic acid, 4-methyl-5-oxazolecarboxylic acid, 5-oxazolecarboxylic acid, 4-methyl, 4-methyl-oxazole-5-carboxylic acid, ksc205g3h, 4-methyl-5-oxazolecarboxylicacid, 4-methyl-oxazol-5-carboxylic acid, 4-methoxyoxazole-5-carboxylic acid, 5-oxazolecarboxylicacid, 4-methyl
PubChem CID 292311
IUPAC Name 4-methyl-1,3-oxazole-5-carboxylic acid
SMILES CC1=C(OC=N1)C(=O)O

Specifications

Melting Point 242°C
Quantity 200 mg
Formula Weight 127.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyloxazole-5-carboxylic Acid