missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Methylnitrosoamino)-1-(3-pyridinyl)-1-butanone, ≥98.0% (GC), MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 64091-91-4 |
| Physical Form | Neat |
| Linear Formula | C10H13N3O2 |
| MDL Number | MFCD00274580 |
| Synonym | 4-(Methylnitrosamino)-1-(3-pyridinyl)-1-butanone; NNK |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 207.23 |
| Percent Purity | ≥98.0% (GC) |
| Grade | Analytical Standard |