missing translation for 'onlineSavingsMsg'
Learn More

4-Methyldiphenylamine 98.0+%, TCI America™

Catalog No. p-7132576
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11731G 1 g
M11735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11731G Supplier TCI America Supplier No. M11731G
Only null left

Chemical Identifiers

CAS 620-84-8
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.25
MDL Number MFCD00092921
InChI Key AGHYMXKKEXDUTA-UHFFFAOYSA-N
Synonym 4-methyldiphenylamine, benzenamine, 4-methyl-n-phenyl, n-phenyl-p-toluidine, 4-methyl-n-phenyl-aniline, 4-methyl-n-phenylbenzamine, 4-methylphenyl phenylamine, phenyl-p-tolylamine, 4-methyldiphenylamin, phenyl-p-tolyl-amine, n-pnenyl-4-methylaniline
PubChem CID 12109
IUPAC Name 4-methyl-N-phenylaniline
SMILES CC1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Melting Point 90°C
Color White-Yellow
Boiling Point 317°C
Quantity 1 g
Formula Weight 183.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyldiphenylamine