missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. p-7132047
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M019625ML 25 mL
M0196500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M019625ML Supplier TCI America Supplier No. M019625ML
Only null left

Chemical Identifiers

CAS 589-91-3
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00001449,MFCD00064171,MFCD00064170
InChI Key MQWCXKGKQLNYQG-UHFFFAOYSA-N
Synonym 4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol
PubChem CID 11524
IUPAC Name 4-methylcyclohexan-1-ol
SMILES CC1CCC(O)CC1

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 114.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methylcyclohexanol (cis- and trans- mixture)
EINECSNumber (3)-2301
TSCA Yes