accessibility menu, dialog, popup

UserName

CH3OH

CAS RN 589-91-3

IUPAC Name: 4-methylcyclohexan-1-ol
Synonyms: 4-methylcyclohexanol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: MQWCXKGKQLNYQG-UHFFFAOYSA-N
SMILES: CC1CCC(O)CC1
Percent Purity 
  • (2)
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)