missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylcyclohexanol, cis + trans, 98%
Description
2-Methylcyclohexanol was used to study the effect of organic solvents on epoxide hydrolase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methylcyclohexanol was used to study the effect of organic solvents on epoxide hydrolase.
Solubility
Soluble in water (15 mg/ml at 20°C), ether, alcohols, and many organic solvents.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid anhydrides, acid chlorides.
2-Methylcyclohexanol was used to study the effect of organic solvents on epoxide hydrolase.
Solubility
Soluble in water (15 mg/ml at 20°C), ether, alcohols, and many organic solvents.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid anhydrides, acid chlorides.
Specifications
Specifications
| Melting Point | -41°C |
| Density | 0.915 |
| Boiling Point | 171°C to 173°C |
| Flash Point | 70°C (158°F) |
| Refractive Index | 1.458 |
| Quantity | 250 mL |
| Beilstein | 1445998 |
| Solubility Information | Soluble in water (15mg/ml at 20°C),ether,alcohols,and many organic solvents. |
| Formula Weight | 114.19 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H227-H302+H312+H332-H315-H320-H335 |
| Hazard Statement | GHS H Statement H227 Combustible liquid. |
| Precautionary Statement | P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P374-P501c |
| EINECSNumber | 209-664-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only