missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcyclohexanol, cis + trans, 98%

Catalog No. AAA16734AU
Change view
Click to view available options
Quantity:
50 mL
250 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA16734AU 1000 mL
AAA16734AD 50 mL
AAA16734AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA16734AU Supplier Thermo Scientific Chemicals Supplier No. A16734AU
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 589-91-3 | C7H14O | 114.19 g/mol

2-Methylcyclohexanol was used to study the effect of organic solvents on epoxide hydrolase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclohexanol was used to study the effect of organic solvents on epoxide hydrolase.

Solubility
Soluble in water (15 mg/ml at 20°C), ether, alcohols, and many organic solvents.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid anhydrides, acid chlorides.

Chemical Identifiers

CAS 589-91-3
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00001449,MFCD00064171,MFCD00064170
InChI Key MQWCXKGKQLNYQG-UHFFFAOYSA-N
Synonym 4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol
PubChem CID 11524
IUPAC Name 4-methylcyclohexan-1-ol
SMILES CC1CCC(O)CC1

Specifications

Melting Point -41°C
Density 0.915
Boiling Point 171°C to 173°C
Flash Point 70°C (158°F)
Refractive Index 1.458
Quantity 1000 mL
Beilstein 1445998
Solubility Information Soluble in water (15mg/ml at 20°C),ether,alcohols,and many organic solvents.
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material 4-Methylcyclohexanol, cis + trans
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H320-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P374-P501c
EINECSNumber 209-664-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only