missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcyclohexanol, 99%, mixture of cis and trans

Catalog No. p-161930
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC166902500 250 g Glass bottle
AC166900010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC166902500 Supplier Thermo Scientific Chemicals Supplier No. 166902500
Only null left

CAS: 589-91-3 | C7H14O | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 589-91-3
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00001449,MFCD00064171,MFCD00064170
InChI Key MQWCXKGKQLNYQG-UHFFFAOYSA-N
Synonym 4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol
PubChem CID 11524
IUPAC Name 4-methylcyclohexan-1-ol
SMILES CC1CCC(O)CC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -41.0°C
Color Colorless to Yellow
Density 0.9200g/mL
Boiling Point 171.0°C to 173.0°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4570 to 1.4590
Linear Formula CH3C6H10OH
Quantity 250 g
Beilstein 06, 14
Specific Gravity 0.92
Solubility Information Solubility in water: slightly soluble
Formula Weight 114.19
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material 4-Methylcyclohexanol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protect
EINECSNumber 209-664-8
TSCA TSCA

RUO – Research Use Only