missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcinnamic acid, predominantly trans, 99%

CAS: 1866-39-3 | C10H10O2 | 162.188 g/mol

$56.33 - $328.34

Chemical Identifiers

CAS 1866-39-3
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00002697
InChI Key RURHILYUWQEGOS-VOTSOKGWSA-N
Synonym 4-methylcinnamic acid, 3-p-tolyl acrylic acid, e-3-p-tolyl acrylic acid, p-methylcinnamic acid, p-methyl cinnamic acid, 2e-3-4-methylphenyl acrylic acid, 2e-3-4-methylphenyl prop-2-enoic acid, 4-methyl-cinnamic acid, e-3-4-methylphenyl-2-propenoic acid, 3-4-methylphenyl-2-propenoic acid
PubChem CID 731767
IUPAC Name (E)-3-(4-methylphenyl)prop-2-enoic acid
SMILES CC1=CC=C(C=C1)C=CC(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1333706
View Documents
Thermo Scientific Chemicals
A1333706
5 g
Each for $56.33
Only null left
 
AAA1333714
View Documents
Thermo Scientific Chemicals
A1333714
25 g
Each for $150.58
Only null left
 
AAA1333722
View Documents
Thermo Scientific Chemicals
A1333722
100 g
Each for $328.34
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1866-39-3
162.188
RURHILYUWQEGOS-VOTSOKGWSA-N
731767
CC1=CC=C(C=C1)C=CC(=O)O
C10H10O2
MFCD00002697
4-methylcinnamic acid, 3-p-tolyl acrylic acid, e-3-p-tolyl acrylic acid, p-methylcinnamic acid, p-methyl cinnamic acid, 2e-3-4-methylphenyl acrylic acid, 2e-3-4-methylphenyl prop-2-enoic acid, 4-methyl-cinnamic acid, e-3-4-methylphenyl-2-propenoic acid, 3-4-methylphenyl-2-propenoic acid
(E)-3-(4-methylphenyl)prop-2-enoic acid

Specifications

1866-39-3
C10H10O2
5 g
4-methylcinnamic acid, 3-p-tolyl acrylic acid, e-3-p-tolyl acrylic acid, p-methylcinnamic acid, p-methyl cinnamic acid, 2e-3-4-methylphenyl acrylic acid, 2e-3-4-methylphenyl prop-2-enoic acid, 4-methyl-cinnamic acid, e-3-4-methylphenyl-2-propenoic acid, 3-4-methylphenyl-2-propenoic acid
CC1=CC=C(C=C1)C=CC(=O)O
162.188
162.19
4-Methylcinnamic acid, predominantly trans
196°C to 200°C
MFCD00002697
1364327
RURHILYUWQEGOS-VOTSOKGWSA-N
(E)-3-(4-methylphenyl)prop-2-enoic acid
731767
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 217-479-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only