missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylbiphenyl 98.0+%, TCI America™
Supplier: TCI America M056325G
Specifications
| 4-Methylbiphenyl | |
| 47°C | |
| 268°C | |
| MFCD00008544 | |
| 4-methylbiphenyl, 4-phenyltoluene, 4-methyl-1,1'-biphenyl, p-methylbiphenyl, p-methyldiphenyl, 4-methyldiphenyl, 1,1'-biphenyl, 4-methyl, biphenyl, 4-methyl, p-phenyltoluene, 4-methylphenyl benzene | |
| CC1=CC=C(C=C1)C1=CC=CC=C1 | |
| 168.24 | |
| 168.24 | |
| Crystalline Powder |
| 644-08-6 | |
| White-Yellow | |
| C13H12 | |
| 25 g | |
| ZZLCFHIKESPLTH-UHFFFAOYSA-N | |
| 4-methyl-1,1'-biphenyl | |
| 12566 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 644-08-6 | |
| 168.24 | |
| ZZLCFHIKESPLTH-UHFFFAOYSA-N | |
| 12566 | |
| CC1=CC=C(C=C1)C1=CC=CC=C1 |
| C13H12 | |
| MFCD00008544 | |
| 4-methylbiphenyl, 4-phenyltoluene, 4-methyl-1,1'-biphenyl, p-methylbiphenyl, p-methyldiphenyl, 4-methyldiphenyl, 1,1'-biphenyl, 4-methyl, biphenyl, 4-methyl, p-phenyltoluene, 4-methylphenyl benzene | |
| 4-methyl-1,1'-biphenyl |
Safety and Handling
EINECSNumber : (4)-0015
RTECSNumber : DV5460000
TSCA : Yes