missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbiphenyl 98.0+%, TCI America™

Catalog No. p-7132240
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M05635G 5 g
M056325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M05635G Supplier TCI America Supplier No. M05635G
Only null left

Chemical Identifiers

CAS 644-08-6
Molecular Formula C13H12
Molecular Weight (g/mol) 168.24
MDL Number MFCD00008544
InChI Key ZZLCFHIKESPLTH-UHFFFAOYSA-N
Synonym 4-methylbiphenyl, 4-phenyltoluene, 4-methyl-1,1'-biphenyl, p-methylbiphenyl, p-methyldiphenyl, 4-methyldiphenyl, 1,1'-biphenyl, 4-methyl, biphenyl, 4-methyl, p-phenyltoluene, 4-methylphenyl benzene
PubChem CID 12566
IUPAC Name 4-methyl-1,1'-biphenyl
SMILES CC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 47°C
Color White-Yellow
Boiling Point 268°C
Quantity 5 g
Formula Weight 168.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylbiphenyl
EINECSNumber (4)-0015
RTECSNumber DV5460000
TSCA Yes