missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzylsulfonyl chloride, 97%

Catalog No. AC454840010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454840010 1 g
AC454840050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC454840010 Supplier Thermo Scientific Chemicals Supplier No. 454840010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 51419-59-1 | C8H9ClO2S | 204.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51419-59-1
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.68
InChI Key JALKUHLLMWYIAT-UHFFFAOYSA-N
Synonym 4-methylbenzylsulfonyl chloride, 4-methylphenyl methanesulfonyl chloride, p-tolylmethanesulfonyl chloride, p-tolyl-methanesulfonyl chloride, p-tolyl methanesulfonyl chloride, 4-methylphenyl methyl sulfonyl chloride, pubchem5592, pubchem5631, acmc-209wgt, 4-methylbenzylsulfonylchloride
PubChem CID 2794648
IUPAC Name (4-methylphenyl)methanesulfonyl chloride
SMILES CC1=CC=C(C=C1)CS(=O)(=O)Cl

Specifications

Melting Point 78.0°C to 82.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 204.68
Percent Purity 97%
Chemical Name or Material 4-Methylbenzylsulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only