CAS RN 51419-59-1
IUPAC Name:
(4-methylphenyl)methanesulfonyl chloride
Synonyms:
(4-methylphenyl)methanesulfonyl chloride
Molecular Weight (g/mol):
204.67
Molecular Formula:
C8H9ClO2S
InChi Key:
JALKUHLLMWYIAT-UHFFFAOYSA-N
SMILES:
CC1=CC=C(CS(Cl)(=O)=O)C=C1
1
–
1
of
1
results
1
Promotions Available
4-Methylbenzylsulfonyl chloride, 97%
4-Methylbenzylsulfonyl chloride, 97%
CAS: 51419-59-1 Molecular Formula: C8H9ClO2S Molecular Weight (g/mol): 204.68 InChI Key: JALKUHLLMWYIAT-UHFFFAOYSA-N Synonym: 4-methylbenzylsulfonyl chloride,4-methylphenyl methanesulfonyl chloride,p-tolylmethanesulfonyl chloride,p-tolyl-methanesulfonyl chloride,p-tolyl methanesulfonyl chloride,4-methylphenyl methyl sulfonyl chloride,pubchem5592,pubchem5631,acmc-209wgt,4-methylbenzylsulfonylchloride PubChem CID: 2794648 IUPAC Name: (4-methylphenyl)methanesulfonyl chloride SMILES: CC1=CC=C(C=C1)CS(=O)(=O)Cl