missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzyl chloride, 98%

Catalog No. AAA1718330
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1718330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1718330 Supplier Thermo Scientific Chemicals Supplier No. A1718330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 104-82-5 | C8H9Cl | 140.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-82-5
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000919
InChI Key DMHZDOTYAVHSEH-UHFFFAOYSA-N
Synonym 4-methylbenzyl chloride, p-xylyl chloride, 1-chloromethyl-4-methylbenzene, alpha-chloro-p-xylene, p-methylbenzyl chloride, benzene, 1-chloromethyl-4-methyl, p-chloromethyl toluene, 4-chloromethyl toluene, p-tolylmethyl chloride, p-xylene, alpha-chloro
PubChem CID 7722
IUPAC Name 1-(chloromethyl)-4-methylbenzene
SMILES CC1=CC=C(C=C1)CCl

Specifications

Melting Point 4°C to 6°C
Density 1.064
Boiling Point 199°C to 201°C
Flash Point 75°C (167°F)
Odor Pungent
Refractive Index 1.533
Quantity 250 g
UN Number UN3265
Beilstein 507387
Merck Index 14,10091
Formula Weight 140.61
Percent Purity 98%
Chemical Name or Material 4-Methylbenzyl chloride
Show More Show Less
Hazard Category H227-H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 203-241-1
RTECSNumber ZE4030000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only