missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzophenone, 98%

Catalog No. p-7022615
Change view
Click to view available options
Quantity:
50 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1547918 50 g
AAA1547936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1547918 Supplier Thermo Scientific Chemicals Supplier No. A1547918

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 134-84-9 | C14H12O | 196.249 g/mol

4-Methylbenzophenone is used as clear coatings for wood, plastic and metal, overprint varnish.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylbenzophenone is used as clear coatings for wood, plastic and metal, overprint varnish.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place. Store in a tightly closed container. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 134-84-9
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.249
MDL Number MFCD00008553
InChI Key WXPWZZHELZEVPO-UHFFFAOYSA-N
Synonym 4-methylbenzophenone, phenyl p-tolyl methanone, 4-methylphenyl phenyl methanone, p-methylbenzophenone, methanone, 4-methylphenyl phenyl, p-benzoyltoluene, 4-methyl benzophenone, phenyl p-tolyl ketone, benzophenone, 4-methyl, p-benzophenone, methyl
PubChem CID 8652
IUPAC Name (4-methylphenyl)-phenylmethanone
SMILES CC1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 53°C to 57°C
Boiling Point 326°C
Flash Point 143°C (289°F)
Quantity 50 g
Beilstein 1909310
Merck Index 14,7317
Solubility Information Insoluble in water.
Formula Weight 196.25
Percent Purity 98%
Chemical Name or Material 4-Methylbenzophenone
EINECSNumber 205-159-1
RTECSNumber DJ1750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only