missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzophenone, 97%

Catalog No. p-159667
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC126350500 50 g Plastic bottle
AC126352500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC126350500 Supplier Thermo Scientific Chemicals Supplier No. 126350500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 134-84-9 | C14H12O | 196.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 134-84-9
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00008553
InChI Key WXPWZZHELZEVPO-UHFFFAOYSA-N
Synonym 4-methylbenzophenone, phenyl p-tolyl methanone, 4-methylphenyl phenyl methanone, p-methylbenzophenone, methanone, 4-methylphenyl phenyl, p-benzoyltoluene, 4-methyl benzophenone, phenyl p-tolyl ketone, benzophenone, 4-methyl, p-benzophenone, methyl
PubChem CID 8652
IUPAC Name (4-methylphenyl)-phenylmethanone
SMILES CC1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 53°C to 57°C
Color Beige to White
Boiling Point 326°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula CH3C6H4COC6H5
Quantity 50 g
Beilstein 07, 440
Merck Index 14, 7317
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and chloroform
Formula Weight 196.25
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Methylbenzophenone, 97%
Show More Show Less
EINECSNumber 205-159-1
RTECSNumber DJ1750000
TSCA TSCA

RUO – Research Use Only