missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylbenzohydrazide 98.0+%, TCI America™
Supplier: TCI America M284625G
Specifications
| 4-Methylbenzohydrazide | |
| 116°C | |
| C8H10N2O | |
| 25 g | |
| MFFVZXOPRXMVET-UHFFFAOYSA-N | |
| 4-methylbenzohydrazide | |
| 77174 | |
| ≥98.0% (GC,T) |
| 3619-22-5 | |
| White-Yellow | |
| MFCD00007607 | |
| p-toluic hydrazide, p-toluic acid hydrazide, p-toluoylhydrazine, 4-methylbenzoylhydrazine, p-toluic acid, hydrazide, 4-methylbenzoic acid hydrazide, p-methylbenzohydrazide, p-toluohydrazide, p-methylbenzoylhydrazine, 4-toluic acid hydrazide | |
| CC1=CC=C(C=C1)C(=O)NN | |
| 150.18 | |
| 150.18 | |
| Crystalline Powder |
Chemical Identifiers
| 3619-22-5 | |
| 150.18 | |
| MFFVZXOPRXMVET-UHFFFAOYSA-N | |
| 77174 | |
| CC1=CC=C(C=C1)C(=O)NN |
| C8H10N2O | |
| MFCD00007607 | |
| p-toluic hydrazide, p-toluic acid hydrazide, p-toluoylhydrazine, 4-methylbenzoylhydrazine, p-toluic acid, hydrazide, 4-methylbenzoic acid hydrazide, p-methylbenzohydrazide, p-toluohydrazide, p-methylbenzoylhydrazine, 4-toluic acid hydrazide | |
| 4-methylbenzohydrazide |
Safety and Handling
TSCA : No