missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzohydrazide 98.0+%, TCI America™

Catalog No. M28465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28465G 5 g
M284625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28465G Supplier TCI America Supplier No. M28465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3619-22-5
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.18
MDL Number MFCD00007607
InChI Key MFFVZXOPRXMVET-UHFFFAOYSA-N
Synonym p-toluic hydrazide, p-toluic acid hydrazide, p-toluoylhydrazine, 4-methylbenzoylhydrazine, p-toluic acid, hydrazide, 4-methylbenzoic acid hydrazide, p-methylbenzohydrazide, p-toluohydrazide, p-methylbenzoylhydrazine, 4-toluic acid hydrazide
PubChem CID 77174
IUPAC Name 4-methylbenzohydrazide
SMILES CC1=CC=C(C=C1)C(=O)NN

Specifications

Melting Point 116°C
Color White-Yellow
Quantity 5 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylbenzohydrazide