missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzenesulfonhydrazide, 97%

Catalog No. p-3758072
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157860250 25 g Plastic bottle
AC157861000 100 g Plastic bottle
AC157865000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC157860250 Supplier Thermo Scientific Chemicals Supplier No. 157860250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1576-35-8 | C7H10N2O2S | 186.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1576-35-8
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00007588
InChI Key ICGLPKIVTVWCFT-UHFFFAOYSA-N
Synonym 4-methylbenzenesulfonhydrazide, p-toluenesulfonhydrazide, tosylhydrazine, p-toluenesulfonyl hydrazide, tosylhydrazide, p-tosylhydrazine, p-toluenesulfonylhydrazine, celogen tsh, ptsh, 4-toluenesulfonyl hydrazide
PubChem CID 15303
SMILES CC1=CC=C(C=C1)S(=O)(=O)NN

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 109.0°C to 113.0°C
Color Gray to White or White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula CH3C6H4SO2NHNH2
Quantity 25 g
Beilstein 11, II, 66
Fieser 01,1185; 02,417; 03,293; 04,511; 05,678; 06,598; 07,375; 08,489; 09,472
Merck Index 15, 9694
Solubility Information Solubility in water: 5g/L (15°C)
IUPAC Name 4-methylbenzenesulfonohydrazide
Formula Weight 186.23
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Methylbenzenesulfonhydrazide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Heating may cause a fire.
Causes serious eye irritation.
Risk of explosion if heated under confinement.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/ combustible materials.
Keep only in orig
EINECSNumber 216-407-3

RUO – Research Use Only