missing translation for 'onlineSavingsMsg'
Learn More

4-Methylanisole, 99%

Catalog No. p-7081555
Change view
Click to view available options
Quantity:
500 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA11572AP 500 mL
AAA115720F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA11572AP Supplier Thermo Scientific Chemicals Supplier No. A11572AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 104-93-8 | C8H10O | 122.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-93-8
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00008413
InChI Key CHLICZRVGGXEOD-UHFFFAOYSA-N
Synonym 4-methylanisole, p-methylanisole, 4-methoxytoluene, p-methoxytoluene, benzene, 1-methoxy-4-methyl, p-cresyl methyl ether, p-cresol methyl ether, anisole, p-methyl, methyl p-tolyl ether, p-tolyl methyl ether
PubChem CID 7731
IUPAC Name 1-methoxy-4-methylbenzene
SMILES CC1=CC=C(C=C1)OC

Specifications

Melting Point -50°C
Density 0.968
Boiling Point 174°C to 175°C
Flash Point 58°C (136°F)
Odor Aromatic
Refractive Index 1.511
Quantity 500 mL
UN Number UN3271
Beilstein 1237336
Formula Weight 122.17
Percent Purity 99%
Chemical Name or Material 4-Methylanisole
Show More Show Less
Hazard Category H226-H302-H315-H319-H361d
Hazard Statement GHS H Statement
H226-H302-H315
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P264b-P270-P280i-P281-P301+P312-P303+P361+P353-P305+P351+P338-P308+P313-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 203-253-7
RTECSNumber BZ8780000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only