missing translation for 'onlineSavingsMsg'
Learn More

4-Methylanisole, 99%, Thermo Scientific Chemicals

Catalog No. p-3779026
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164662500 250 g Glass bottle
AC164660010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC164662500 Supplier Thermo Scientific Chemicals Supplier No. 164662500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 104-93-8 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-93-8
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00008413
InChI Key CHLICZRVGGXEOD-UHFFFAOYSA-N
Synonym 4-methylanisole, p-methylanisole, 4-methoxytoluene, p-methoxytoluene, benzene, 1-methoxy-4-methyl, p-cresyl methyl ether, p-cresol methyl ether, anisole, p-methyl, methyl p-tolyl ether, p-tolyl methyl ether
PubChem CID 7731
IUPAC Name 1-methoxy-4-methylbenzene
SMILES CC1=CC=C(C=C1)OC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -50°C
Color Colorless to Yellow
Density 0.9600g/mL
Boiling Point 174°C
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5100 to 1.513
Linear Formula CH3C6H4OCH3
Quantity 250 g
Beilstein 06, 392
Specific Gravity 0.96
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alchol
Formula Weight 122.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Methylanisole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Suspected of damaging the unborn child.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
EINECSNumber 203-253-7

RUO – Research Use Only