missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylamino)pyridine 98.0+%, TCI America™

$44.73 - $173.37

Chemical Identifiers

CAS 1121-58-0
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00152625
InChI Key LSCYTCMNCWMCQE-UHFFFAOYSA-N
Synonym 4-methylamino pyridine, 4-methylaminopyridine, n-methyl-4-pyridinamine, 4-pyridinamine, n-methyl, pyridin-4-yl methyl amine, 4-mamp, dmap on polystyrene, pubchem12915, pyridin-4yl-methylamine, pyridin-4-yl-methylamine
PubChem CID 123098
IUPAC Name N-methylpyridin-4-amine
SMILES CNC1=CC=NC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M21831G
View Documents
TCI America
M21831G
1 g
Each for $44.73
Only null left
 
M21835G
View Documents
TCI America
M21835G
5 g
Each for $173.37
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1121-58-0
108.14
LSCYTCMNCWMCQE-UHFFFAOYSA-N
123098
CNC1=CC=NC=C1
C6H8N2
MFCD00152625
4-methylamino pyridine, 4-methylaminopyridine, n-methyl-4-pyridinamine, 4-pyridinamine, n-methyl, pyridin-4-yl methyl amine, 4-mamp, dmap on polystyrene, pubchem12915, pyridin-4yl-methylamine, pyridin-4-yl-methylamine
N-methylpyridin-4-amine

Specifications

1121-58-0
White-Yellow
C6H8N2
1 g
LSCYTCMNCWMCQE-UHFFFAOYSA-N
N-methylpyridin-4-amine
123098
≥98.0% (GC,T)
4-(Methylamino)pyridine
127°C
100°C
MFCD00152625
4-methylamino pyridine, 4-methylaminopyridine, n-methyl-4-pyridinamine, 4-pyridinamine, n-methyl, pyridin-4-yl methyl amine, 4-mamp, dmap on polystyrene, pubchem12915, pyridin-4yl-methylamine, pyridin-4-yl-methylamine
CNC1=CC=NC=C1
108.14
108.14
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents