missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Methylamino)pyridine 98.0+%, TCI America™
Supplier: TCI America M21835G
Specifications
| 4-(Methylamino)pyridine | |
| 127°C | |
| 100°C | |
| MFCD00152625 | |
| 4-methylamino pyridine, 4-methylaminopyridine, n-methyl-4-pyridinamine, 4-pyridinamine, n-methyl, pyridin-4-yl methyl amine, 4-mamp, dmap on polystyrene, pubchem12915, pyridin-4yl-methylamine, pyridin-4-yl-methylamine | |
| CNC1=CC=NC=C1 | |
| 108.14 | |
| 108.14 | |
| Crystalline Powder |
| 1121-58-0 | |
| White-Yellow | |
| C6H8N2 | |
| 5 g | |
| LSCYTCMNCWMCQE-UHFFFAOYSA-N | |
| N-methylpyridin-4-amine | |
| 123098 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 1121-58-0 | |
| 108.14 | |
| LSCYTCMNCWMCQE-UHFFFAOYSA-N | |
| 123098 | |
| CNC1=CC=NC=C1 |
| C6H8N2 | |
| MFCD00152625 | |
| 4-methylamino pyridine, 4-methylaminopyridine, n-methyl-4-pyridinamine, 4-pyridinamine, n-methyl, pyridin-4-yl methyl amine, 4-mamp, dmap on polystyrene, pubchem12915, pyridin-4yl-methylamine, pyridin-4-yl-methylamine | |
| N-methylpyridin-4-amine |
Safety and Handling
TSCA : Yes