missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylamino)phenol Sulfate 98.0+%, TCI America™

Catalog No. p-7132016
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M014525G 25 g
M0145500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M014525G Supplier TCI America Supplier No. M014525G
Only null left

Chemical Identifiers

CAS 55-55-0
Molecular Formula C7H9NO
MDL Number MFCD00013140

Specifications

Melting Point 260°C
Color Gray-White
Quantity 25 g
UN Number 3077
Formula Weight 344.38
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylamino)phenol Sulfate
EINECSNumber (3)-0676
RTECSNumber SL8650000
TSCA Yes