CAS RN 55-55-0
IUPAC Name:
bis(4-(methylamino)phenol); sulfuric acid
Synonyms:
metol p-methylaminophenolsulfate paramethylaminophenol sulfate N-methyl-p-aminophenol sulfate p-methylaminophenol sulfate 4-methylaminophenol sulfate 4-(methylamino)phenol sulfate
Molecular Weight (g/mol):
344.38
Molecular Formula:
C14H20N2O6S
InChi Key:
ZVNPWFOVUDMGRP-UHFFFAOYSA-N
SMILES:
OS(O)(=O)=O.CNC1=CC=C(O)C=C1.CNC1=CC=C(O)C=C1
1
–
4
of
4
results
1
Promotions Available
4-(Methylamino)phenol sulfate, 99%
4-(Methylamino)phenol sulfate, 99%
CAS: 55-55-0 Molecular Formula: C14H18N2O2·H2SO4 MDL Number: MFCD00013140 Synonym: Bis(4-hydroxy-N-methylanilinium) sulfate; Metol
2
4-(Methylamino)phenol sulfate, ACS, 99.0-101.5%
CAS: 55-55-0 MDL Number: MFCD00013140 Synonym: Bis(4-hydroxy-n-methylanilinium) sulfate; Metol
3
Promotions Available
4-Methylaminophenol sulfate, 99%, extra pure
4-Methylaminophenol sulfate, 99%, extra pure
CAS: 55-55-0 Molecular Formula: C14H18N2O2·H2SO4 Molecular Weight (g/mol): 344.38 MDL Number: MFCD00013140 Synonym: Metol
4
4-(Methylamino)phenol Sulfate 98.0+%, TCI America™
CAS: 55-55-0 Molecular Formula: C7H9NO MDL Number: MFCD00013140