missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Methylamino)benzoic Acid 98.0+%, TCI America™
Supplier: TCI America M026725G
Specifications
| 4-(Methylamino)benzoic Acid | |
| 161°C | |
| C8H9NO2 | |
| 25 g | |
| ZVIDMSBTYRSMAR-UHFFFAOYSA-N | |
| 4-(methylamino)benzoic acid | |
| 66345 | |
| 151.17 | |
| Crystalline Powder |
| 10541-83-0 | |
| White-Yellow | |
| MFCD00002535 | |
| 4-methylamino benzoic acid, benzoic acid, 4-methylamino, p-methylaminobenzoic acid, n-methyl-4-aminobenzoic acid, 4-n-methylamino benzoic acid, n-methyl-4-aminobenzoate, benzoic acid, p-methylamino, 4-methylamino-benzoic acid, p-methylamino benzoic acid, p-n-methylamino benzoic acid | |
| CNC1=CC=C(C=C1)C(=O)O | |
| 151.165 | |
| CHEBI:7308 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 10541-83-0 | |
| 151.165 | |
| ZVIDMSBTYRSMAR-UHFFFAOYSA-N | |
| 66345 | |
| 4-(methylamino)benzoic acid |
| C8H9NO2 | |
| MFCD00002535 | |
| 4-methylamino benzoic acid, benzoic acid, 4-methylamino, p-methylaminobenzoic acid, n-methyl-4-aminobenzoic acid, 4-n-methylamino benzoic acid, n-methyl-4-aminobenzoate, benzoic acid, p-methylamino, 4-methylamino-benzoic acid, p-methylamino benzoic acid, p-n-methylamino benzoic acid | |
| CHEBI:7308 | |
| CNC1=CC=C(C=C1)C(=O)O |
Safety and Handling
EINECSNumber : (3)-1449
TSCA : Yes