missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylamino)benzoic Acid 98.0+%, TCI America™

Catalog No. M02675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M02675G 5 g
M026725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M02675G Supplier TCI America Supplier No. M02675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10541-83-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00002535
InChI Key ZVIDMSBTYRSMAR-UHFFFAOYSA-N
Synonym 4-methylamino benzoic acid, benzoic acid, 4-methylamino, p-methylaminobenzoic acid, n-methyl-4-aminobenzoic acid, 4-n-methylamino benzoic acid, n-methyl-4-aminobenzoate, benzoic acid, p-methylamino, 4-methylamino-benzoic acid, p-methylamino benzoic acid, p-n-methylamino benzoic acid
PubChem CID 66345
ChEBI CHEBI:7308
IUPAC Name 4-(methylamino)benzoic acid
SMILES CNC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 161°C
Color White-Yellow
Quantity 5 g
Formula Weight 151.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylamino)benzoic Acid
EINECSNumber (3)-1449
TSCA Yes