missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylamino)benzoic acid, 97%

CAS: 10541-83-0 | C8H9NO2 | 151.165 g/mol

$66.92 - $803.22

Chemical Identifiers

CAS 10541-83-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00002535
InChI Key ZVIDMSBTYRSMAR-UHFFFAOYSA-N
Synonym 4-methylamino benzoic acid, benzoic acid, 4-methylamino, p-methylaminobenzoic acid, n-methyl-4-aminobenzoic acid, 4-n-methylamino benzoic acid, n-methyl-4-aminobenzoate, benzoic acid, p-methylamino, 4-methylamino-benzoic acid, p-methylamino benzoic acid, p-n-methylamino benzoic acid
PubChem CID 66345
ChEBI CHEBI:7308
IUPAC Name 4-(methylamino)benzoic acid
SMILES CNC1=CC=C(C=C1)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1242606
View Documents
Thermo Scientific Chemicals
A1242606
5 g
Each for $66.92
Only null left
 
AAA1242614
View Documents
Thermo Scientific Chemicals
A1242614
25 g
Each for $224.19
Only null left
 
AAA1242622
View Documents
Thermo Scientific Chemicals
A1242622
100 g
Each for $803.22
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-(Methylamino)benzoic acid important for the preparation of other pharmaceutical products, dyes, flavors, and preservatives. .For the preparation of medicines and other fine chemicals.

Solubility
Slightly soluble. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

10541-83-0
151.165
ZVIDMSBTYRSMAR-UHFFFAOYSA-N
66345
4-(methylamino)benzoic acid
C8H9NO2
MFCD00002535
4-methylamino benzoic acid, benzoic acid, 4-methylamino, p-methylaminobenzoic acid, n-methyl-4-aminobenzoic acid, 4-n-methylamino benzoic acid, n-methyl-4-aminobenzoate, benzoic acid, p-methylamino, 4-methylamino-benzoic acid, p-methylamino benzoic acid, p-n-methylamino benzoic acid
CHEBI:7308
CNC1=CC=C(C=C1)C(=O)O

Specifications

10541-83-0
C8H9NO2
5 g
4-methylamino benzoic acid, benzoic acid, 4-methylamino, p-methylaminobenzoic acid, n-methyl-4-aminobenzoic acid, 4-n-methylamino benzoic acid, n-methyl-4-aminobenzoate, benzoic acid, p-methylamino, 4-methylamino-benzoic acid, p-methylamino benzoic acid, p-n-methylamino benzoic acid
ZVIDMSBTYRSMAR-UHFFFAOYSA-N
4-(methylamino)benzoic acid
66345
151.17
4- (Methylamino)benzoic acid
158°C to 160°C
MFCD00002535
2082805
Slightly soluble. Soluble in alcohol.
CNC1=CC=C(C=C1)C(=O)O
151.165
CHEBI:7308
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 234-121-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only