missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylamino)benzoic acid, 97%

Catalog No. AAA1242606
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1242606 5 g
AAA1242614 25 g
AAA1242622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1242606 Supplier Thermo Scientific Chemicals Supplier No. A1242606

CAS: 10541-83-0 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-(Methylamino)benzoic acid important for the preparation of other pharmaceutical products, dyes, flavors, and preservatives. .For the preparation of medicines and other fine chemicals.

Solubility
Slightly soluble. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 10541-83-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00002535
InChI Key ZVIDMSBTYRSMAR-UHFFFAOYSA-N
Synonym 4-methylamino benzoic acid, benzoic acid, 4-methylamino, p-methylaminobenzoic acid, n-methyl-4-aminobenzoic acid, 4-n-methylamino benzoic acid, n-methyl-4-aminobenzoate, benzoic acid, p-methylamino, 4-methylamino-benzoic acid, p-methylamino benzoic acid, p-n-methylamino benzoic acid
PubChem CID 66345
ChEBI CHEBI:7308
IUPAC Name 4-(methylamino)benzoic acid
SMILES CNC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 158°C to 160°C
Quantity 5 g
Beilstein 2082805
Solubility Information Slightly soluble. Soluble in alcohol.
Formula Weight 151.17
Percent Purity 97%
Chemical Name or Material 4- (Methylamino)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 234-121-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only