missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Methylacetophenone 95.0+%, TCI America™
$32.72 - $152.08
Chemical Identifiers
| CAS | 122-00-9 |
|---|---|
| Molecular Formula | C9H10O |
| Molecular Weight (g/mol) | 134.18 |
| MDL Number | MFCD00008751 |
| InChI Key | GNKZMNRKLCTJAY-UHFFFAOYSA-N |
| Synonym | 4'-methylacetophenone, p-methylacetophenone, 1-p-tolyl ethanone, 1-p-tolylethanone, melilotal, 4-methylacetophenone, 4-acetyltoluene, 1-4-methylphenyl ethanone, methyl p-tolyl ketone, p-acetotoluene |
| PubChem CID | 8500 |
| IUPAC Name | 1-(4-methylphenyl)ethan-1-one |
| SMILES | CC(=O)C1=CC=C(C)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M013525ML
|
TCI America
M013525ML |
25 mL |
Each for $32.72
|
|
|||||
|
M0135500ML
|
TCI America
M0135500ML |
500 mL |
Each for $152.08
|
|
|||||
Chemical Identifiers
| 122-00-9 | |
| 134.18 | |
| GNKZMNRKLCTJAY-UHFFFAOYSA-N | |
| 8500 | |
| CC(=O)C1=CC=C(C)C=C1 |
| C9H10O | |
| MFCD00008751 | |
| 4'-methylacetophenone, p-methylacetophenone, 1-p-tolyl ethanone, 1-p-tolylethanone, melilotal, 4-methylacetophenone, 4-acetyltoluene, 1-4-methylphenyl ethanone, methyl p-tolyl ketone, p-acetotoluene | |
| 1-(4-methylphenyl)ethan-1-one |
Specifications
| 122-00-9 | |
| 226°C | |
| MFCD00008751 | |
| 4'-methylacetophenone, p-methylacetophenone, 1-p-tolyl ethanone, 1-p-tolylethanone, melilotal, 4-methylacetophenone, 4-acetyltoluene, 1-4-methylphenyl ethanone, methyl p-tolyl ketone, p-acetotoluene | |
| CC(=O)C1=CC=C(C)C=C1 | |
| 134.18 | |
| 134.18 | |
| Liquid |
| Yellow | |
| C9H10O | |
| 25 mL | |
| GNKZMNRKLCTJAY-UHFFFAOYSA-N | |
| 1-(4-methylphenyl)ethan-1-one | |
| 8500 | |
| ≥95.0% (GC) | |
| 4′-Methylacetophenone |
Safety and Handling
EINECSNumber : (3)-1239
RTECSNumber : AM9463000
TSCA : Yes