accessibility menu, dialog, popup

UserName

H3COCH3

CAS RN 122-00-9

IUPAC Name: 1-(4-methylphenyl)ethan-1-one
Synonyms: P-methylacetophenone P-methyl acetophenone
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: GNKZMNRKLCTJAY-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C)C=C1
Boiling Point 
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)