missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylacetophenone 95.0+%, TCI America™

Catalog No. M013525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M013525ML 25 mL
M0135500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M013525ML Supplier TCI America Supplier No. M013525ML

Chemical Identifiers

CAS 122-00-9
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00008751
InChI Key GNKZMNRKLCTJAY-UHFFFAOYSA-N
Synonym 4'-methylacetophenone, p-methylacetophenone, 1-p-tolyl ethanone, 1-p-tolylethanone, melilotal, 4-methylacetophenone, 4-acetyltoluene, 1-4-methylphenyl ethanone, methyl p-tolyl ketone, p-acetotoluene
PubChem CID 8500
IUPAC Name 1-(4-methylphenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(C)C=C1

Specifications

Color Yellow
Boiling Point 226°C
Quantity 25 mL
Formula Weight 134.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Methylacetophenone
EINECSNumber (3)-1239
RTECSNumber AM9463000
TSCA Yes