missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylacetophenone, 96%

CAS: 122-00-9 | C9H10O | 134.18 g/mol

$48.03 - $180.05

Chemical Identifiers

CAS 122-00-9
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00008751
InChI Key GNKZMNRKLCTJAY-UHFFFAOYSA-N
Synonym 4'-methylacetophenone, p-methylacetophenone, 1-p-tolyl ethanone, 1-p-tolylethanone, melilotal, 4-methylacetophenone, 4-acetyltoluene, 1-4-methylphenyl ethanone, methyl p-tolyl ketone, p-acetotoluene
PubChem CID 8500
IUPAC Name 1-(4-methylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1446918
View Documents
Thermo Scientific Chemicals
A1446918
50 g
Each for $48.03
Only null left
 
AAA1446930
View Documents
Thermo Scientific Chemicals
A1446930
250 g
Each for $72.55
Only null left
 
AAA144690B
View Documents
Thermo Scientific Chemicals
A144690B
1000 g
Each for $180.05
Only null left
 
Description

Description

4'-Methylacetophenone is used as a flavoring agent. It reacts with morpholine to get 4-(p-tolyl-thioacetyl)-morpholine in the presence of sulfur as a reagent. Further, it is used as an intermediate in the manufacture of active pharmaceutical ingredients, perfumes and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Methylacetophenone is used as a flavoring agent. It reacts with morpholine to get 4-(p-tolyl-thioacetyl)-morpholine in the presence of sulfur as a reagent. Further, it is used as an intermediate in the manufacture of active pharmaceutical ingredients, perfumes and cosmetics.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Specifications

Chemical Identifiers

122-00-9
134.18
GNKZMNRKLCTJAY-UHFFFAOYSA-N
8500
CC(=O)C1=CC=C(C)C=C1
C9H10O
MFCD00008751
4'-methylacetophenone, p-methylacetophenone, 1-p-tolyl ethanone, 1-p-tolylethanone, melilotal, 4-methylacetophenone, 4-acetyltoluene, 1-4-methylphenyl ethanone, methyl p-tolyl ketone, p-acetotoluene
1-(4-methylphenyl)ethanone

Specifications

122-00-9
1.01
92°C (197°F)
1.534
50 g
4'-methylacetophenone, p-methylacetophenone, 1-p-tolyl ethanone, 1-p-tolylethanone, melilotal, 4-methylacetophenone, 4-acetyltoluene, 1-4-methylphenyl ethanone, methyl p-tolyl ketone, p-acetotoluene
GNKZMNRKLCTJAY-UHFFFAOYSA-N
1-(4-methylphenyl)ethanone
8500
96%
-19°C
224°C to 226°C
C9H10O
MFCD00008751
606053
Miscible with water.
CC(=O)C1=CC=C(C)C=C1
134.18
134.18
4'-Methylacetophenone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227
Toxic if swallowed.
Combustible liquid.

P210-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P370+P378q-P501c

H227-H302-H315-H319

EINECSNumber : 204-514-8

RTECSNumber : AM9463000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only