missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylacetanilide, 98+%

Catalog No. AAA1372122
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1372122 100 g
AAA1372114 25 g
AAA1372136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1372122 Supplier Thermo Scientific Chemicals Supplier No. A1372122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 103-89-9 | C9H11NO | 149.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103-89-9
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD00008677
InChI Key YICAMJWHIUMFDI-UHFFFAOYSA-N
Synonym 4'-methylacetanilide, p-acetotoluidide, n-4-methylphenyl acetamide, 4-methylacetanilide, n-acetyl-p-toluidine, p-acetotoluide, acetyl-p-toluidine, p-acetotoluidine, acetamide, n-4-methylphenyl, 4-acetotoluide
PubChem CID 7684
IUPAC Name N-(4-methylphenyl)acetamide
SMILES CC1=CC=C(C=C1)NC(=O)C

Specifications

Melting Point 147°C to 150°C
Boiling Point 306°C to 308°C
Quantity 100 g
Beilstein 607036
Merck Index 14,74
Formula Weight 149.19
Percent Purity ≥98%
Chemical Name or Material 4'-Methylacetanilide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 203-155-4
RTECSNumber AN2930000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only