missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl L-Aspartate Hydrochloride 95.0+%, TCI America™
$74.56 - $223.68
Chemical Identifiers
| CAS | 16856-13-6 |
|---|---|
| Molecular Formula | C5H10ClNO4 |
| Molecular Weight (g/mol) | 183.588 |
| MDL Number | MFCD00038972 |
| InChI Key | QRBMPUYOGOCYDJ-DFWYDOINSA-N |
| Synonym | h-asp ome-oh.hcl, l-aspartic acid beta-methyl ester hydrochloride, s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, h-asp ome-oh hcl, 4-methyl l-aspartate hydrochloride, s-2-amino-succinic acid 4-methyl ester hydrochloride, 2s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, l-aspartic acid 4-methyl ester hcl, aspartic acid, 4-methylester, hydrochloride, l-aspartic acid, 4-methyl ester, hydrochloride |
| PubChem CID | 12832742 |
| IUPAC Name | (2S)-2-amino-4-methoxy-4-oxobutanoic acid;hydrochloride |
| SMILES | COC(=O)CC(C(=O)O)N.Cl |
Chemical Identifiers
| 16856-13-6 | |
| 183.588 | |
| QRBMPUYOGOCYDJ-DFWYDOINSA-N | |
| 12832742 | |
| COC(=O)CC(C(=O)O)N.Cl |
| C5H10ClNO4 | |
| MFCD00038972 | |
| h-asp ome-oh.hcl, l-aspartic acid beta-methyl ester hydrochloride, s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, h-asp ome-oh hcl, 4-methyl l-aspartate hydrochloride, s-2-amino-succinic acid 4-methyl ester hydrochloride, 2s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, l-aspartic acid 4-methyl ester hcl, aspartic acid, 4-methylester, hydrochloride, l-aspartic acid, 4-methyl ester, hydrochloride | |
| (2S)-2-amino-4-methoxy-4-oxobutanoic acid;hydrochloride |
Specifications
| 16856-13-6 | |
| White-Yellow | |
| MFCD00038972 | |
| h-asp ome-oh.hcl, l-aspartic acid beta-methyl ester hydrochloride, s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, h-asp ome-oh hcl, 4-methyl l-aspartate hydrochloride, s-2-amino-succinic acid 4-methyl ester hydrochloride, 2s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, l-aspartic acid 4-methyl ester hcl, aspartic acid, 4-methylester, hydrochloride, l-aspartic acid, 4-methyl ester, hydrochloride | |
| COC(=O)CC(C(=O)O)N.Cl | |
| 183.588 | |
| 183.59 | |
| Crystalline Powder |
| 193°C | |
| C5H10ClNO4 | |
| 5 g | |
| QRBMPUYOGOCYDJ-DFWYDOINSA-N | |
| (2S)-2-amino-4-methoxy-4-oxobutanoic acid;hydrochloride | |
| 12832742 | |
| ≥95.0% (T) | |
| 4-Methyl L-Aspartate Hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator