missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl L-Aspartate Hydrochloride 95.0+%, TCI America™

Catalog No. M20245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20245G 5 g
M202425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20245G Supplier TCI America Supplier No. M20245G

Chemical Identifiers

CAS 16856-13-6
Molecular Formula C5H10ClNO4
Molecular Weight (g/mol) 183.588
MDL Number MFCD00038972
InChI Key QRBMPUYOGOCYDJ-DFWYDOINSA-N
Synonym h-asp ome-oh.hcl, l-aspartic acid beta-methyl ester hydrochloride, s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, h-asp ome-oh hcl, 4-methyl l-aspartate hydrochloride, s-2-amino-succinic acid 4-methyl ester hydrochloride, 2s-2-amino-4-methoxy-4-oxobutanoic acid hydrochloride, l-aspartic acid 4-methyl ester hcl, aspartic acid, 4-methylester, hydrochloride, l-aspartic acid, 4-methyl ester, hydrochloride
PubChem CID 12832742
IUPAC Name (2S)-2-amino-4-methoxy-4-oxobutanoic acid;hydrochloride
SMILES COC(=O)CC(C(=O)O)N.Cl

Specifications

Melting Point 193°C
Color White-Yellow
Quantity 5 g
Formula Weight 183.59
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl L-Aspartate Hydrochloride
TSCA No
Recommended Storage Refrigerator