missing translation for 'onlineSavingsMsg'
Learn More

4-(Methyl-d3-nitrosamino)-1-(3-pyridyl)-1-butanol, ≥97% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111017978
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017978 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017978 Supplier Merck Emd Millipore Supplier No. 944541MG

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706031, T706021

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 1020719-61-2
Physical Form Neat
Linear Formula C10D3H12N3O2
MDL Number MFCD03425608
Synonym 4-(Methyl-d3-nitrosoamino)-1-(3-pyridinyl)-1-butanol; NNAL-Methyl-d3
Type Analytical Standard
Quantity 1 mg
Formula Weight 212.26
Percent Purity ≥97% (HPLC)
Grade Analytical Standard