missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-beta-styrylboronic acid pinacol ester, 98%

CAS: 149777-84-4 | C15H21BO2 | 244.141 g/mol

Supplier:  Thermo Scientific Chemicals L1969803

 View more versions of this product

Catalog No. AAL1969803


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Methyl-beta-styrylboronic acid pinacol ester
56°C to 58°C
MFCD03788749
7204616
HHBWKASJNTZJLB-ZHACJKMWSA-N
4,4,5,5-tetramethyl-2-[(E)-2-(4-methylphenyl)ethenyl]-1,3,2-dioxaborolane
11042960
98%
149777-84-4
C15H21BO2
1 g
4-methyl-beta-styrylboronic acid pinacol ester, e-4,4,5,5-tetramethyl-2-4-methylstyryl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-e-2-4-methylphenyl ethenyl-1,3,2-dioxaborolane, 4-methyl-b-styrylboronic acid pinacol ester, 4-methyl-, a-styrylboronic acid pinacol ester, 2-e-4-methylstyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, beta-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl 4-methylstyrene
B1(OC(C(O1)(C)C)(C)C)C=CC2=CC=C(C=C2)C
244.141
244.14

Chemical Identifiers

149777-84-4
244.141
HHBWKASJNTZJLB-ZHACJKMWSA-N
11042960
B1(OC(C(O1)(C)C)(C)C)C=CC2=CC=C(C=C2)C
C15H21BO2
MFCD03788749
4-methyl-beta-styrylboronic acid pinacol ester, e-4,4,5,5-tetramethyl-2-4-methylstyryl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-e-2-4-methylphenyl ethenyl-1,3,2-dioxaborolane, 4-methyl-b-styrylboronic acid pinacol ester, 4-methyl-, a-styrylboronic acid pinacol ester, 2-e-4-methylstyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, beta-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl 4-methylstyrene
4,4,5,5-tetramethyl-2-[(E)-2-(4-methylphenyl)ethenyl]-1,3,2-dioxaborolane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only