missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-6,7-methylenedioxycoumarin, TCI America™

Catalog No. M09711G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M09711G Supplier TCI America Supplier No. M09711G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15071-04-2
Molecular Formula C11H8O4
Molecular Weight (g/mol) 204.181
MDL Number MFCD00143343
InChI Key XAOGHIKJQRXPHX-UHFFFAOYSA-N
Synonym 4-Methylayapin
PubChem CID 609315
IUPAC Name 8-methyl-[1,3]dioxolo[4,5-g]chromen-6-one
SMILES CC1=CC(=O)OC2=CC3=C(C=C12)OCO3

Specifications

Melting Point 181°C
Color White-Yellow
Quantity 1 g
Formula Weight 204.18
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-6,7-methylenedioxycoumarin