accessibility menu, dialog, popup

UserName

CAS RN 15071-04-2

CH3OOOO

CAS RN 15071-04-2

IUPAC Name: 8-methyl-2H,6H-[1,3]dioxolo[4,5-g]chromen-6-one
Synonyms: 8-methyl-2H-[1,3]dioxolo[4,5-g]chromen-6-one
Molecular Weight (g/mol): 204.18
Molecular Formula: C11H8O4
InChi Key: XAOGHIKJQRXPHX-UHFFFAOYSA-N
SMILES: CC1=CC(=O)OC2=CC3=C(OCO3)C=C12
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results