missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-5-imidazolecarboxaldehyde, 99%

Catalog No. AC425770250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425770250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC425770250 Supplier Thermo Scientific Chemicals Supplier No. 425770250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 68282-53-1 | C5H6N2O | 110.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68282-53-1
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00173728
InChI Key KMWCSNCNHSEXIF-UHFFFAOYSA-N
Synonym 4-methyl-1h-imidazole-5-carbaldehyde, 5-methylimidazole-4-carboxaldehyde, 4-methylimidazole-5-carboxaldehyde, 4-methyl-5-imidazolecarboxaldehyde, 5-methyl-3h-imidazole-4-carbaldehyde, 4-methylimidazole-5-carbaldehyde, 1h-imidazole-4-carboxaldehyde, 5-methyl, 1h-imidazole-5-carboxaldehyde, 4-methyl, pubchem23016, acmc-1b6zs
PubChem CID 2795887
IUPAC Name 5-methyl-1H-imidazole-4-carbaldehyde
SMILES CC1=C(C=O)N=CN1

Specifications

Melting Point 165.0°C to 166.0°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 110.12
Percent Purity 99%
Chemical Name or Material 4-Methyl-5-imidazolecarboxaldehyde

RUO – Research Use Only