missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitrobenzoic Acid 99.0+%, TCI America™

Catalog No. M095025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M095025G 25 g
M0950500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M095025G Supplier TCI America Supplier No. M095025G

Chemical Identifiers

CAS 96-98-0
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007174
InChI Key BBEWSMNRCUXQRF-UHFFFAOYSA-N
Synonym 4-methyl-3-nitrobenzoic acid, 3-nitro-4-methylbenzoic acid, 3-nitro-p-toluic acid, benzoic acid, 4-methyl-3-nitro, 3-nitro-para-toluic acid, m-nitro-p-toluic acid, 4-methyl-3-nitro-benzoic acid, p-toluic acid, 3-nitro, 3-nitro-4-methyl benzoic acid, benzoic acid,4-methyl-3-nitro
PubChem CID 7319
SMILES CC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 190°C
Color Yellow
Quantity 25 g
Formula Weight 181.15
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-3-nitrobenzoic Acid
EINECSNumber (3)-1510
TSCA Yes