missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-3-nitrobenzenesulfonyl Chloride 98.0+%, TCI America™
Supplier: TCI America N070425G
Specifications
| 4-Methyl-3-nitrobenzenesulfonyl Chloride | |
| 36°C | |
| C7H6ClNO4S | |
| 25 g | |
| 4-methyl-3-nitrobenzene-1-sulfonyl chloride, 2-nitrotoluene-4-sulfonyl chloride, 4-methyl-3-nitro-benzenesulfonyl chloride, 4-methyl-3-nitrobenzenesulfonylchloride, chloro 4-methyl-3-nitrophenyl sulfone, 4-methyl-3-nitrobenzolsulfonylchlorid, pubchem5674, acmc-1awd3, 3-nitro-p-toluenesulfonyl chloride, 3-nitro-4-methyl phenylsulfonyl chloride | |
| CC1=C(C=C(C=C1)S(=O)(=O)Cl)[N+](=O)[O-] | |
| 235.638 | |
| 235.64 | |
| Crystalline Powder |
| 616-83-1 | |
| 154°C | |
| MFCD00129811 | |
| 3261 | |
| OQFYBGANSUNUAO-UHFFFAOYSA-N | |
| 4-methyl-3-nitrobenzenesulfonyl chloride | |
| 560638 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 616-83-1 | |
| 235.638 | |
| OQFYBGANSUNUAO-UHFFFAOYSA-N | |
| 560638 | |
| CC1=C(C=C(C=C1)S(=O)(=O)Cl)[N+](=O)[O-] |
| C7H6ClNO4S | |
| MFCD00129811 | |
| 4-methyl-3-nitrobenzene-1-sulfonyl chloride, 2-nitrotoluene-4-sulfonyl chloride, 4-methyl-3-nitro-benzenesulfonyl chloride, 4-methyl-3-nitrobenzenesulfonylchloride, chloro 4-methyl-3-nitrophenyl sulfone, 4-methyl-3-nitrobenzolsulfonylchlorid, pubchem5674, acmc-1awd3, 3-nitro-p-toluenesulfonyl chloride, 3-nitro-4-methyl phenylsulfonyl chloride | |
| 4-methyl-3-nitrobenzenesulfonyl chloride |
Safety and Handling
TSCA : No