missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitrobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. N07045G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07045G 5 g
N070425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07045G Supplier TCI America Supplier No. N07045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 616-83-1
Molecular Formula C7H6ClNO4S
Molecular Weight (g/mol) 235.638
MDL Number MFCD00129811
InChI Key OQFYBGANSUNUAO-UHFFFAOYSA-N
Synonym 4-methyl-3-nitrobenzene-1-sulfonyl chloride, 2-nitrotoluene-4-sulfonyl chloride, 4-methyl-3-nitro-benzenesulfonyl chloride, 4-methyl-3-nitrobenzenesulfonylchloride, chloro 4-methyl-3-nitrophenyl sulfone, 4-methyl-3-nitrobenzolsulfonylchlorid, pubchem5674, acmc-1awd3, 3-nitro-p-toluenesulfonyl chloride, 3-nitro-4-methyl phenylsulfonyl chloride
PubChem CID 560638
IUPAC Name 4-methyl-3-nitrobenzenesulfonyl chloride
SMILES CC1=C(C=C(C=C1)S(=O)(=O)Cl)[N+](=O)[O-]

Specifications

Melting Point 36°C
Boiling Point 154°C
Quantity 5 g
UN Number 3261
Formula Weight 235.64
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-3-nitrobenzenesulfonyl Chloride