missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitrobenzenesulfonyl chloride, 95%, Thermo Scientific Chemicals

Catalog No. p-7024739
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1993106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1993106 Supplier Thermo Scientific Chemicals Supplier No. A1993106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 616-83-1 | Molecular Formula: C7H6ClNO4S | Molecular Weight: 235.638 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 616-83-1
Molecular Formula C7H6ClNO4S
Molecular Weight (g/mol) 235.638
MDL Number MFCD00129811
InChI Key OQFYBGANSUNUAO-UHFFFAOYSA-N
Synonym 4-methyl-3-nitrobenzene-1-sulfonyl chloride, 2-nitrotoluene-4-sulfonyl chloride, 4-methyl-3-nitro-benzenesulfonyl chloride, 4-methyl-3-nitrobenzenesulfonylchloride, chloro 4-methyl-3-nitrophenyl sulfone, 4-methyl-3-nitrobenzolsulfonylchlorid, pubchem5674, acmc-1awd3, 3-nitro-p-toluenesulfonyl chloride, 3-nitro-4-methyl phenylsulfonyl chloride
PubChem CID 560638
IUPAC Name 4-methyl-3-nitrobenzenesulfonyl chloride
SMILES CC1=C(C=C(C=C1)S(=O)(=O)Cl)[N+](=O)[O-]

Specifications

Melting Point 33°C to 36°C
Boiling Point 152°C to 154°C (1 mmHg)
Quantity 5 g
UN Number UN3261
Beilstein 2696481
Sensitivity Moisture sensitive
Formula Weight 235.65
Percent Purity 95%
Chemical Name or Material 4-Methyl-3-nitrobenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only