missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitrobenzeneboronic acid, 98%

Catalog No. AAL1705206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1705206 5 g
AAL1705203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1705206 Supplier Thermo Scientific Chemicals Supplier No. L1705206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 80500-27-2 | C7H8BNO4 | 180.954 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80500-27-2
Molecular Formula C7H8BNO4
Molecular Weight (g/mol) 180.954
MDL Number MFCD00191550
InChI Key OASVXBRTNVFKFS-UHFFFAOYSA-N
Synonym 4-methyl-3-nitrophenyl boronic acid, 4-methyl-3-nitrobenzeneboronic acid, 3-nitro-p-tolylboronic acid, 4-methyl-3-nitrophenyl boranediol, 4-methyl-3-nitro-phenyl boronic acid, 3-nitro-4-methylphenylboronic acid, 2-nitrotoluene-4-boronic acid, boronic acid, 4-methyl-3-nitrophenyl, 3-nitro-4-methylbenzeneboronic acid
PubChem CID 2773515
IUPAC Name (4-methyl-3-nitrophenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)C)[N+](=O)[O-])(O)O

Specifications

Melting Point ∼250°C
Quantity 5 g
Beilstein 3279018
Formula Weight 180.96
Percent Purity 98%
Chemical Name or Material 4-Methyl-3-nitrobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only