missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitroaniline, 98%

CAS: 119-32-4 | C7H8N2O2 | 152.153 g/mol

$135.40 - $402.49

Chemical Identifiers

CAS 119-32-4
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00007910
InChI Key GDIIPKWHAQGCJF-UHFFFAOYSA-N
Synonym 4-amino-2-nitrotoluene, 3-nitro-4-methylaniline, 3-nitro-p-toluidine, m-nitro-p-toluidine, 2-nitro-4-aminotoluene, benzenamine, 4-methyl-3-nitro, 5-nitro-4-toluidine, p-toluidine, 3-nitro, gl-amin czech, gl-amin
PubChem CID 8390
ChEBI CHEBI:81670
IUPAC Name 4-methyl-3-nitroaniline
SMILES CC1=C(C=C(C=C1)N)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1253814
View Documents
Thermo Scientific Chemicals
A1253814
25 g
Each for $135.40
Only null left
 
AAA1253822
View Documents
Thermo Scientific Chemicals
A1253822
100 g
Each for $402.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

119-32-4
152.153
GDIIPKWHAQGCJF-UHFFFAOYSA-N
8390
4-methyl-3-nitroaniline
C7H8N2O2
MFCD00007910
4-amino-2-nitrotoluene, 3-nitro-4-methylaniline, 3-nitro-p-toluidine, m-nitro-p-toluidine, 2-nitro-4-aminotoluene, benzenamine, 4-methyl-3-nitro, 5-nitro-4-toluidine, p-toluidine, 3-nitro, gl-amin czech, gl-amin
CHEBI:81670
CC1=C(C=C(C=C1)N)[N+](=O)[O-]

Specifications

119-32-4
1.312
Characteristic
MFCD00007910
UN2660
4-amino-2-nitrotoluene, 3-nitro-4-methylaniline, 3-nitro-p-toluidine, m-nitro-p-toluidine, 2-nitro-4-aminotoluene, benzenamine, 4-methyl-3-nitro, 5-nitro-4-toluidine, p-toluidine, 3-nitro, gl-amin czech, gl-amin
CC1=C(C=C(C=C1)N)[N+](=O)[O-]
152.153
CHEBI:81670
98%
74°C to 79°C
175°C (347°F)
C7H8N2O2
25 g
638984
GDIIPKWHAQGCJF-UHFFFAOYSA-N
4-methyl-3-nitroaniline
8390
152.15
4-Methyl-3-nitroaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P314-P501c

H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROTOLUIDINES (MONO)

EINECSNumber : 204-314-0

RTECSNumber : XU8227000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only