missing translation for 'onlineSavingsMsg'
Learn More

(4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazin-7-yl)methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC62009CB

 View more versions of this product

Catalog No. CC62009CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

(4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazin-7-yl)methanol
96.5
97%
C9H12N2O2
250mg
PPBFQXGUORHMBA-UHFFFAOYSA-N
(4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazin-7-yl)methanol
24229648
97%
921938-81-0
100.0
Amber Glass Bottle
MFCD09817507
4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methanol, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazin-7-yl methanol, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazin-7-yl methan-1-ol, 2h-pyrido 3,2-b-1,4-oxazine-7-methanol,3,4-dihydro-4-methyl
CN1CCOC2=C1N=CC(=C2)CO
180.207
180.21

Chemical Identifiers

921938-81-0
180.207
PPBFQXGUORHMBA-UHFFFAOYSA-N
24229648
CN1CCOC2=C1N=CC(=C2)CO
C9H12N2O2
MFCD09817507
4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methanol, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazin-7-yl methanol, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazin-7-yl methan-1-ol, 2h-pyrido 3,2-b-1,4-oxazine-7-methanol,3,4-dihydro-4-methyl
(4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazin-7-yl)methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions