missing translation for 'onlineSavingsMsg'
Learn More

(4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazin-7-yl)methanol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 921938-81-0
Molecular Formula C9H12N2O2
Molecular Weight (g/mol) 180.207
MDL Number MFCD09817507
InChI Key PPBFQXGUORHMBA-UHFFFAOYSA-N
Synonym 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methanol, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazin-7-yl methanol, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazin-7-yl methan-1-ol, 2h-pyrido 3,2-b-1,4-oxazine-7-methanol,3,4-dihydro-4-methyl
PubChem CID 24229648
IUPAC Name (4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazin-7-yl)methanol
SMILES CN1CCOC2=C1N=CC(=C2)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC62009CB
SDS View Documents
Thermo Scientific Chemicals
CC62009CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC62009DA
SDS View Documents
Thermo Scientific Chemicals
CC62009DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

921938-81-0
180.207
PPBFQXGUORHMBA-UHFFFAOYSA-N
24229648
CN1CCOC2=C1N=CC(=C2)CO
C9H12N2O2
MFCD09817507
4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methanol, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazin-7-yl methanol, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazin-7-yl methan-1-ol, 2h-pyrido 3,2-b-1,4-oxazine-7-methanol,3,4-dihydro-4-methyl
(4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazin-7-yl)methanol

Specifications

921938-81-0
100.0
Amber Glass Bottle
MFCD09817507
4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methanol, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazin-7-yl methanol, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazin-7-yl methan-1-ol, 2h-pyrido 3,2-b-1,4-oxazine-7-methanol,3,4-dihydro-4-methyl
CN1CCOC2=C1N=CC(=C2)CO
180.207
180.21
(4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazin-7-yl)methanol
96.5
97%
C9H12N2O2
250mg
PPBFQXGUORHMBA-UHFFFAOYSA-N
(4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazin-7-yl)methanol
24229648
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents