missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-(trifluoromethyl)benzonitrile, 98%

Catalog No. AAH3190406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3190406 5 g
AAH3190403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3190406 Supplier Thermo Scientific Chemicals Supplier No. H3190406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 261952-05-0 | C9H6F3N | 185.15 g/mol

It is used as primary and secondary reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as primary and secondary reagent.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 261952-05-0
Molecular Formula C9H6F3N
Molecular Weight (g/mol) 185.15
MDL Number MFCD01631596
InChI Key WCZWEODOIUKVHX-UHFFFAOYSA-N
Synonym 4-methyl-2-trifluoromethyl benzonitrile, 4-methyl-2-trifluoromethyl benzenecarbonitrile, 2-cyano-5-methylbenzotrifluoride, 2-trifluoromethyl-p-tolunitrile, 4-cyano-3-trifluoromethyl toluene, benzonitrile,4-methyl-2-trifluoromethyl
PubChem CID 2775597
IUPAC Name 4-methyl-2-(trifluoromethyl)benzonitrile
SMILES CC1=CC(=C(C=C1)C#N)C(F)(F)F

Specifications

Melting Point 22°C to 28°C
Quantity 5 g
UN Number UN3439
Solubility Information Insoluble in water.
Formula Weight 185.15
Percent Purity 98%
Chemical Name or Material 4-Methyl-2-(trifluoromethyl)benzonitrile
Hazard Category H302+H332-H311-H315-H319
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only